Thermo Scientific Chemicals

3-Chlorophenol, 98+%, Thermo Scientific Chemicals

Catalog number: A13671.30
250 g, Each
Thermo Scientific Chemicals

3-Chlorophenol, 98+%, Thermo Scientific Chemicals

Catalog number: A13671.30
250 g, Each
Quantity
Catalog number: A13671.30
also known as A13671-30
Price (USD)
Quantity
-

Chemical Identifiers

CAS
108-43-0
IUPAC Name
3-chlorophenol
Molecular Formula
C6H5ClO
InChI Key
HORNXRXVQWOLPJ-UHFFFAOYSA-N
SMILES
OC1=CC=CC(Cl)=C1
Assay (GC)
≥97.5%
Appearance (Color)
White to yellow to green to brown or green/brown
Refractive Index
1.5650 to 1.5670 @ 20?C
Form
Fused solid or clear liquid as melt

Description

3-Chlorophenol is widely used in the pharmaceutical, dyes and organic synthesis industry. It is an important intermediate in the synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorophenol is widely used in the pharmaceutical, dyes and organic synthesis industry. It is also used as a disinfectant. It is an important intermediate in the synthesis of chemicals.

Solubility
Soluble in ethanol and ether.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text