2-Isobutyl-3-methoxypyrazine, 98%
2-Isobutyl-3-methoxypyrazine, 98%
2-Isobutyl-3-methoxypyrazine, 98%
2-Isobutyl-3-methoxypyrazine, 98%
Thermo Scientific Chemicals

2-Isobutyl-3-methoxypyrazine, 98%

CAS: 24683-00-9 | C9H14N2O | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A13680.06
also known as A13680-06
Price (USD)
180.65
Online Exclusive
200.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
180.65
Online Exclusive
200.00
Save 19.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS24683-00-9
IUPAC Name2-methoxy-3-(2-methylpropyl)pyrazine
Molecular FormulaC9H14N2O
InChI KeyUXFSPRAGHGMRSQ-UHFFFAOYSA-N
SMILESCOC1=NC=CN=C1CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4895-1.4935 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only