CTS™ Neurobasal™ Medium
Gibco™

CTS™ Neurobasal™ Medium

CTS Neurobasal Medium is a regulatory-compliant medium for neural stem cell and neuron growth, intended for regenerative medicine applications.
Have Questions?
Catalog NumberQuantity
A1371201500 mL
Catalog number A1371201
Price (KRW)
157,000
Online offer
Ends: 31-Dec-2026
174,000
Save 17,000
Each
Quantity:
500 mL
Price (KRW)
157,000
Online offer
Ends: 31-Dec-2026
174,000
Save 17,000
Each

CTS Neurobasal Medium is a chemically defined medium specifically designed to support the growth and viability of pure populations of neural stem cells and PSC-derived neurons. Leveraging a classic formulation trusted by neuroscientists for over 30 years, this medium enhances neuronal survival and differentiation. The CTS version adheres to stringent quality standards and includes comprehensive regulatory documentation, making it ideal for cell therapy manufacturing and clinical research applications.

Benefits of CTS Neurobasal Medium include:

  • Enhanced neuronal support—proven to support the survival and differentiation of neurons
  • Serum-free—eliminates the need for serum or an astrocyte feeder layer when used with CTS B-27 Supplement XenoFree
  • High-quality standards—manufactured at FDA-registered sites that conform to cGMP for medical devices (21 CFR Part 820) and are ISO 13485 and ISO 9001 certified
  • Regulatory compliance—comes with specific intended-use statements, full documentation traceability, and easy access to our Drug Master File (DMF)

Why choose Gibco CTS products?

Gibco CTS products are designed to provide you with a proven choice for transitioning your cell therapy to the clinic with confidence. Our products come with the necessary documentation to support your regulatory filings, ensuring a smooth and compliant process.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell TypeNeural Cells
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
FormatBottle
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
Product TypeCell Culture Medium
Quantity500 mL
Shelf Life12 months from date of manufacture
Shipping ConditionAmbient
ClassificationAnimal Origin-free
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C and protect from light.

Frequently asked questions (FAQs)

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.