5-Nonanone, 98%
5-Nonanone, 98%
5-Nonanone, 98%
Thermo Scientific Chemicals

5-Nonanone, 98%

CAS: 502-56-7 | C9H18O | 142.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A13722.22
also known as A13722-22
Price (EUR)
60,80
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
60,80
Each
Add to cart
Chemical Identifiers
CAS502-56-7
IUPAC Namenonan-5-one
Molecular FormulaC9H18O
InChI KeyWSGCRAOTEDLMFQ-UHFFFAOYSA-N
SMILESCCCCC(=O)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4180-1.4220 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only