cis-2-Hexen-1-ol, 94%, remainder mainly trans isomer
cis-2-Hexen-1-ol, 94%, remainder mainly trans isomer
cis-2-Hexen-1-ol, 94%, remainder mainly trans isomer
cis-2-Hexen-1-ol, 94%, remainder mainly trans isomer
Thermo Scientific Chemicals

cis-2-Hexen-1-ol, 94%, remainder mainly trans isomer

CAS: 928-94-9 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13734.06
also known as A13734-06
Price (EUR)
64,90
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
64,90
Each
Add to cart
Chemical Identifiers
CAS928-94-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥92.5% (major isomer)
Refractive Index1.4365-1.4435 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only