5-Acetyl-2,4-dimethylthiazole, 99%
5-Acetyl-2,4-dimethylthiazole, 99%
5-Acetyl-2,4-dimethylthiazole, 99%
Thermo Scientific Chemicals

5-Acetyl-2,4-dimethylthiazole, 99%

CAS: 38205-60-6 | C7H9NOS | 155.215 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13741.06
also known as A13741-06
Price (EUR)
21,90
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
21,90
Each
Chemical Identifiers
CAS38205-60-6
IUPAC Name1-(2,4-dimethyl-1,3-thiazol-5-yl)ethan-1-one
Molecular FormulaC7H9NOS
InChI KeyBLQOKWQUTLNKON-UHFFFAOYSA-N
SMILESCC(=O)C1=C(C)N=C(C)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Infrared spectrum1.5395-1.5445 @ 20?C
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only