1-Chloro-2,4-dinitrobenzene, 98%
1-Chloro-2,4-dinitrobenzene, 98%
1-Chloro-2,4-dinitrobenzene, 98%
Thermo Scientific Chemicals

1-Chloro-2,4-dinitrobenzene, 98%

CAS: 97-00-7 | C6H3ClN2O4 | 202.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A13774.0E
also known as A13774-0E
Price (EUR)
330,00
Each
Quantity:
2500 g
Request bulk or custom format
Price (EUR)
330,00
Each
Chemical Identifiers
CAS97-00-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow to orange to brown to black
FormCrystals or powder or crystalline powder or lumps or fused solid or flakes or powder and chunks
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)46.0-53.0°C
1-Chloro-2,4-dinitrobenzene is used as a reagent for the detection and determination of pyridine compounds. It has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH). It is an irreversible inhibitor of human thioredoxin reductase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2,4-dinitrobenzene is used as a reagent for the detection and determination of pyridine compounds. It has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH). It is an irreversible inhibitor of human thioredoxin reductase.

Solubility
Soluble in chloroform, ether, hot alcohol, and benzene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, ammonia. Reacts with hydrazine hydrate.
RUO – Research Use Only

General References:

  1. Elias S. J. Arner; Mikael Bjornstedt and Arne Holmgren. 1-Chloro-2,4-dinitrobenzene Is an Irreversible Inhibitor of Human Thioredoxin Reductase. Loss of thioredoxin disulfide reductase activity is accompanied by a large increase in nadph oxidase activity.Journal of Biological Chemistry.1995, 270 3479-3482.
  2. YC Awasthi; HS Garg; DD Dao; CA Partridge and SK Srivastava. Enzymatic conjugation of erythrocyte glutathione with 1-chloro-2,4-dinitrobenzene: the fate of glutathione conjugate in erythrocytes and the effect of glutathione depletion on hemoglobin. bloodjournal.hematologylibrary.1981, 58 733-738.
  3. The doubly-activated chloro-substituent is readily displaced by nucleophiles, e.g.: Ammonia/ammonium acetate to give 2,4-dinitroaniline: Org. Synth. Coll., 2, 221 (1943); or with ammonia gas in toluene in the presence of TBAB which improves the solubility of ammonia in the organic medium and accelerates the reaction: J. Chem. Soc., Chem. Commun., 1267 (1987).
  4. Benzyl mercaptan to give the thioether: Org. Synth. Coll., 5, 474 (1973).
  5. Iodide ion to give 2,4-dinitro-1-iodobenzene: Org. Synth. Coll., 5, 478 (1973). Nitrite in toluene (phase-transfer) to give 1,2,4-trinitrobenzene (caution!): J. Org. Chem., 56, 4967 (1991).