Ethyl acetopyruvate, 98%
Ethyl acetopyruvate, 98%
Ethyl acetopyruvate, 98%
Thermo Scientific Chemicals

Ethyl acetopyruvate, 98%

CAS: 615-79-2 | C7H10O4 | 158.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13808.06
also known as A13808-06
Price (EUR)
52,40
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
52,40
Each
Chemical Identifiers
CAS615-79-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4720-1.4760 @ 20°C
Ethyl acetopyruvate is used in the preparation of 5-methyl-isoxazole-3-carboxylic acid ethyl ester. It is involved in the preparation of ethyl 5-amino-2-methylpyndine-4-carboxyIate by reacting with nitroacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl acetopyruvate is used in the preparation of 5-methyl-isoxazole-3-carboxylic acid ethyl ester. It is involved in the preparation of ethyl 5-amino-2-methylpyndine-4-carboxyIate by reacting with nitroacetamide.

Solubility
Miscible with water, ether,chloroform, methanol and ethanol.

Notes
Hygroscopic. Store in a cool place. Incompatible with acids, bases, oxidizing agents and reducing agents.
RUO – Research Use Only

General References:

  1. Cegłowski, M.; Schroeder, G. Removal of heavy metal ions with the use of chelating polymers obtained by grafting pyridine-pyrazole ligands onto polymethylhydrosiloxane. Chem. Eng. J. 2015, 259, 885-893.
  2. Kasare, S.; Bankar, S. K.; Ramasastry, S. S. V. Expeditious Metal-Free Access to Functionalized Polycyclic Acetals under Mild Aqueous Conditions. Org. Lett. 2014, 16 (16), 4284-4287.