6-Amino-2-methylquinoline, 97%
6-Amino-2-methylquinoline, 97%
6-Amino-2-methylquinoline, 97%
6-Amino-2-methylquinoline, 97%
Thermo Scientific Chemicals

6-Amino-2-methylquinoline, 97%

CAS: 65079-19-8 | C10H10N2 | 158.204 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A13967.03
also known as A13967-03
Price (TWD)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS65079-19-8
IUPAC Name2-methylquinolin-6-amine
Molecular FormulaC10H10N2
InChI KeyTYJFYUVDUUACKX-UHFFFAOYSA-N
SMILESCC1=CC=C2C=C(N)C=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)182.0-192.0?C
Appearance (Color)Brown
FormPowder
6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Hitoshi Kusama; Hironori Arakawa. Influence of quinoline derivatives in I-/I3- redox electrolyte solution on the performance of Ru(II)-dye-sensitized nanocrystalline TiO2 solar cell. Journal of Photochemistry and Photobiology A: Chemistry. 2004, 165,(1-3), 157-163.
  2. Shannon M.Biros; Julius Rebek, Jr. Structure and binding properties of water-soluble cavitands and capsules. Chem. Soc. Rev. 2007, 36,(1), 93-104.