4-Chlorobenzylamine, 97+%
4-Chlorobenzylamine, 97+%
4-Chlorobenzylamine, 97+%
4-Chlorobenzylamine, 97+%
Thermo Scientific Chemicals

4-Chlorobenzylamine, 97+%

CAS: 104-86-9 | C7H8ClN | 141.598 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13984.06
also known as A13984-06
Price (TWD)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS104-86-9
IUPAC Name1-(4-chlorophenyl)methanamine
Molecular FormulaC7H8ClN
InChI KeyYMVFJGSXZNNUDW-UHFFFAOYSA-N
SMILESNCC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.0%
Refractive Index1.5565-1.5615 @ 20?C
Identification (FTIR)Conforms
FormLiquid
4-Chlorobenzylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzylamine is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
RUO – Research Use Only

General References:

  1. F.Markwardt; H.Landmann; P.Walsmann. Comparative studies on the inhibition of trypsin, plasmin, and thrombin by derivatives of benzylamine and benzamidine. European Journal of Biochemistry. 1968, 6,(4), 502-506.
  2. Robin S. Eichen Conn; Alan W.Douglas; Sandor Karady; Edward G.Corley; Alfred V.Lovell; Ichiro Shinkai. An unusual Fischer indole synthesis with 4-keto acids: an indole incorporating the terminal hydrazine nitrogen. J. Org. Chem. 1990, 55,(9), 2908-2913.