5-Chlorosalicylaldehyde, 98%
5-Chlorosalicylaldehyde, 98%
5-Chlorosalicylaldehyde, 98%
5-Chlorosalicylaldehyde, 98%
Thermo Scientific Chemicals

5-Chlorosalicylaldehyde, 98%

CAS: 635-93-8 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14069.14
also known as A14069-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS635-93-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Melting Point (clear melt)99.0-105.0?C
FormPowder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only