1-Bromo-2,4-difluorobenzene, 98+%
1-Bromo-2,4-difluorobenzene, 98+%
1-Bromo-2,4-difluorobenzene, 98+%
Thermo Scientific Chemicals

1-Bromo-2,4-difluorobenzene, 98+%

CAS: 348-57-2 | C6H3BrF2 | 192.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14098.36
also known as A14098-36
Price (EUR)
479,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
479,00
Each
Chemical Identifiers
CAS348-57-2
IUPAC Name1-bromo-2,4-difluorobenzene
Molecular FormulaC6H3BrF2
InChI KeyMGHBDQZXPCTTIH-UHFFFAOYSA-N
SMILESFC1=CC=C(Br)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.5035-1.5075 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only