Piperonylic acid, 98+%
Piperonylic acid, 98+%
Piperonylic acid, 98+%
Thermo Scientific Chemicals

Piperonylic acid, 98+%

CAS: 94-53-1 | C8H6O4 | 166.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A14105.22
also known as A14105-22
Price (EUR)
195,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
195,00
Each
Chemical Identifiers
CAS94-53-1
IUPAC Name2H-1,3-benzodioxole-5-carboxylic acid
Molecular FormulaC8H6O4
InChI KeyVDVJGIYXDVPQLP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0% to ≤102.0%
Assay (Silylated GC)≥98.0%
Identification (FTIR)Conforms
Appearance (Color)White to yellow or cream to pale brown
Piperonylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperonylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Michel Schalk; Francisco Cabello-Hurtado; Marie-Agnes Pierrel; Rossitza Atanassova; Patrick Saindrenan; Daniele Werck-Reichhart. Piperonylic acid, a selective, mechanism-based inactivator of the trans-cinnamate 4-hydroxylase: A new tool to control the flux of metabolites in the phenylpropanoid pathway. Plant Physiology. 1998, 118,(1), 209-218.
  2. Esaac G.Esaac; John E.Casida. Piperonylic acid conjugates with alanine, glutamate, glutamine, glycine, and serine in living houseflies. Journal of Insect Physiology. 1968, 14,(7), 920-925.