4,4'-Dibromobiphenyl, 98+%
4,4'-Dibromobiphenyl, 98+%
4,4'-Dibromobiphenyl, 98+%
Thermo Scientific Chemicals

4,4'-Dibromobiphenyl, 98+%

CAS: 92-86-4 | C12H8Br2 | 312.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A14139.30
also known as A14139-30
Price (EUR)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS92-86-4
IUPAC Name4,4'-dibromo-1,1'-biphenyl
Molecular FormulaC12H8Br2
InChI KeyHQJQYILBCQPYBI-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)163.0-169.0°C
Appearance (Color)White to cream to yellow to pale brown
FormCrystals or powder or crystalline powder
4,4'-Dibromobiphenyl is commonly used as fire retardants and plasticizers. It is also have application in gas chromatography and liquid chromatography analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dibromobiphenyl is commonly used as fire retardants and plasticizers. It is also have application in gas chromatography and liquid chromatography analysis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Sierra Rayne; Michael G Ikonomou; MacMurray D Whale. Photoelectrocatalytic degradation of 4,4'-dibromobiphenyl in aqueous solution on TiO2and doped TiO2nanotube arrays. Chemosphere. 2011, 82 (1), 43-47.
  2. Sierra Rayne; Michael G Ikonomou; MacMurray D Whale. Anaerobic microbial and photochemical degradation of 4,4'-dibromodiphenyl ether. Water Research. 2003, 37 (3), 551-560.