Thermo Scientific Chemicals

4-n-Hexylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14168.06
5 g, Each
Thermo Scientific Chemicals

4-n-Hexylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14168.06
5 g, Each
Quantity
Catalog number: A14168.06
also known as A14168-06
Price (USD)
Price: 203.00
Online price: 175.65
Your price:
Quantity
-

Chemical Identifiers

CAS
21643-38-9
IUPAC Name
4-hexylbenzoic acid
Molecular Formula
C13H18O2
InChI Key
CPEPWESLFZVUEP-UHFFFAOYSA-N
SMILES
CCCCCCC1=CC=C(C=C1)C(O)=O
Form
Powder or flakes
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.5% to ≤101.5%
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.5% to ≤101.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text