TargetSeq™ Oligo Blocker P1

Catalog number: A14178

Applied Biosystems™  Related applications: DNA Sequencing | SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

TargetSeq™Oligo Blockers are designed to enable multiplexed library enrichment with all TargetSeq™Enrichment kits. The blocker oligonucleotides function to prevent cross-hybridization of the library adaptors during the solution-phase DNA probe capture step of the target enrichment protocol. TargetSeq™Oligo Blocker P1 is a single blocker oligo specific for the P1 sequencing adaptor and must be used in combination with TargetSeq™Oligo Blocker 1-16. These blocker oligos are sold individually but used together during the library capture in order to block the adapters at both ends of the library molecules. TargetSeq™Oligo Blocker P1is compatible with SOLiD® 4 and 5500 genetic analyzer fragment libraries that use the P1 sequencing adaptor and P2 adaptor barcodes 1-16.
• Optimized blocker design to reduce unwanted adaptor cross-hybridization during target enrichment for your 5500 Genetic Analyzer pipeline
• Conveniently provided at ready-to-use concentrations for fast reaction set-up and optimized performance
• Pre-mixed and validated to provide end users with significant time and cost savings

Optimized Blocker Designs
TargetSeq™Oligo Blocker P1 is a single blocker oligo specific for the P1 library adapter that hs been validated to provide optimal performance during multiplexed (4-plex) sample enrichment using full length barcoded libraries. Typical on-target read percentages are 70-85%, indicating that the blocker oligos are effectively preventing adaptor cross-hybridization during capture.

Convenience of Ready-to-Use Solution
TargetSeq™Oligo Blockers are optimized to provide exceptional and economical performance when combined with SOLiD® barcoded libraries (indexes 1-16) and SOLiD® 4 or 5500 Series genetic analysis systems. They diluted and ready to be added to capture reactions making experimental setup fast and easy. TargetSeq™enrichment systems are one of the first products lines to endorse multiplex sample capture (4-plex) using library barcoding and pre-capture sample pooling which dramatically lowers the per sample costs and simplifies the workflow.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Blocker Oligos
Key Function: Sequencing Target Enrichment (Block Cross-Hybridization)
For Use With (Equipment): 5500 Genetic Analyzer, SOLiD™ 4 System
Product Line: TargetSeq™
Product Size: 24 reactions
Shipping Condition: Dry Ice

Contents & storage

Store at -15°C to -25°C

Documents

Manuals & protocols