2,4,5-Trifluorobenzoic acid, 98%
2,4,5-Trifluorobenzoic acid, 98%
2,4,5-Trifluorobenzoic acid, 98%
2,4,5-Trifluorobenzoic acid, 98%
Thermo Scientific Chemicals

2,4,5-Trifluorobenzoic acid, 98%

CAS: 446-17-3 | C7H2F3O2 | 175.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14193.06
also known as A14193-06
Price (EUR)
56,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
56,00
Each
Add to cart
Chemical Identifiers
CAS446-17-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)97.0-103.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only