3-Iodoanisole, 98%
3-Iodoanisole, 98%
3-Iodoanisole, 98%
Thermo Scientific Chemicals

3-Iodoanisole, 98%

CAS: 766-85-8 | C7H7IO | 234.036 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14208.06
also known as A14208-06
Price (EUR)
29,90
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
29,90
Each
Chemical Identifiers
CAS766-85-8
IUPAC Name1-iodo-3-methoxybenzene
Molecular FormulaC7H7IO
InChI KeyRSHBAGGASAJQCH-UHFFFAOYSA-N
SMILESCOC1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Refractive Index1.6110-1.6150 @20?C
FormLiquid
3-Iodoanisole is used to produce 3,3'-Dimethoxy-biphenyl. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodoanisole is used to produce 3,3′-Dimethoxy-biphenyl. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in chloroform, alcohol, ether. Insoluble in water.

Notes
Light Sensitive. Incompatible with light and oxidizing agents. Store in the dark.
RUO – Research Use Only
Elaine Crosbie; Alan R Kennedy; Robert E Mulvey; Stuart D Robertson. After-effects of lithium-mediated alumination of 3-iodoanisole: isolation of molecular salt elimination and trapped-benzyne products. Dalton Transactions (Print Edition).2012, 41 (6), 1832-1839.