Benzeneboronic acid, 98+%
Benzeneboronic acid, 98+%
Benzeneboronic acid, 98+%
Thermo Scientific Chemicals

Benzeneboronic acid, 98+%

CAS: 98-80-6 | C6H7BO2 | 121.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14257.18
also known as A14257-18
Price (EUR)
161,65
Online Exclusive
179,00
Save 17,35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
161,65
Online Exclusive
179,00
Save 17,35 (10%)
Each
Chemical Identifiers
CAS98-80-6
IUPAC Namephenylboronic acid
Molecular FormulaC6H7BO2
InChI KeyHXITXNWTGFUOAU-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0%
Assay (HPLC)≥98.0%
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Benzeneboronic acid is widely utilized in various cross coupling reactions for example Mizoroki-Heck and Suzuki-Miyaura coupling reactions, etc. It serves as a receptor for carbohydrates, antimicrobial agents and enzyme inhibitors in biological reactions. It acts as a reagent in palladium-catalyzed stereoselective Heck-type reaction and as a catalyst in Ni(II) pincer complex and Pd(II) pyridoxal hydrazone metallacycles. It is also employed as a sensor for neutron capture therapy for cancer, transmembrane transport and bioconjugation and labeling of proteins and cell surface.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Avoid moisture exposure.
RUO – Research Use Only

General References:

  1. Reagent for protection of diols as their cyclic boronates: J. Am. Chem. Soc., 80, 2443 (1958); Tetrahedron, 25, 477 (1969), which are also useful in GC and GC-MS. This has been utilized to trap cis-diols, in order to prevent over-oxidation during dihydroxylation reactions catalyzed by Osmium(VIII) oxide, 12103: Chem. Lett., 1721 (1988); J. Org. Chem., 63, 7322 (1998).
  2. Promotes the ortho-hydroxalkylation of phenols by aldehydes. The cyclic boronate, formed via a [3,3] sigmatropic rearrangement, is the key intermediate: Synthesis, 365 (1979):
  3. With citronellal, hexahydrocannabinoids are formed: J. Chem. Soc., Perkin 1, 605 (1992).
  4. Acts as a template for Diels-Alder reactions, by forming boronate linkages with a hydroxydiene and a hydroxydienophile: Synthesis 1171 (1991); for Nicolaou's application to synthesis of the CD ring system of taxol, see: J. Chem. Soc., Chem. Commun., 1118 (1992); J. Am. Chem. Soc., 117, 634 (1995):
  5. For use in the formation of an oxazaborolidine catalyst for use in enantioselective reductions, see note under (S)-(-)-ɑ,ɑ-Diphenyl prolinol, L09217.
  6. Forms a stable chiral acyloxyborane (CAB) catalyst with tartaric acid derivatives, which catalyze hetero Diels-Alder reactions, e.g. between aldehydes and 1-Methoxy-3-trimethyl siloxy-1,3-butadiene, L06100, to give enantioselectively, dihydro-4-pyrone derivatives: Tetrahedron, 50, 979 (1994).
  7. The most widely-used reaction of arylboronic acids is the Pd-catalyzed Suzuki synthesis of unsymmetrical biaryls: Synth. Commun., 11, 513 (1981):
  8. For illustrative example, see: Org. Synth., 75, 53 (1997).
  9. For a brief feature on uses of this reagent in Organic synthesis, see: Synlett, 2679 (2006). For further information and reviews on boronic acid chemistry, see Appendix 5.