2,3,4-Trimethoxybenzoic acid, 98+%
2,3,4-Trimethoxybenzoic acid, 98+%
2,3,4-Trimethoxybenzoic acid, 98+%
Thermo Scientific Chemicals

2,3,4-Trimethoxybenzoic acid, 98+%

CAS: 573-11-5 | C10H12O5 | 212.201 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14278.18
also known as A14278-18
Price (TWD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS573-11-5
IUPAC Name2,3,4-trimethoxybenzoate
Molecular FormulaC10H11O5
InChI KeyHZNQSWJZTWOTKM-UHFFFAOYSA-M
SMILESCOC1=CC=C(C([O-])=O)C(OC)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
Melting Point (clear melt)98.0-104.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only