4-Bromo-3-methylbenzoic acid, 98%
4-Bromo-3-methylbenzoic acid, 98%
4-Bromo-3-methylbenzoic acid, 98%
4-Bromo-3-methylbenzoic acid, 98%
Thermo Scientific Chemicals

4-Bromo-3-methylbenzoic acid, 98%

CAS: 7697-28-1 | C8H6BrO2 | 214.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14302.06
also known as A14302-06
Price (EUR)
72,40
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
72,40
Each
Add to cart
Chemical Identifiers
CAS7697-28-1
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)211.0-218.0?C
Appearance (Color)White to cream to pale orange
Assay (HPLC)≥97.5%
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only