5,6-Dicloro-2-mercaptobencimidazol, 98 %, Thermo Scientific Chemicals
5,6-Dicloro-2-mercaptobencimidazol, 98 %, Thermo Scientific Chemicals
5,6-Dicloro-2-mercaptobencimidazol, 98 %, Thermo Scientific Chemicals
5,6-Dicloro-2-mercaptobencimidazol, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5,6-Dicloro-2-mercaptobencimidazol, 98 %, Thermo Scientific Chemicals

CAS: 19462-98-7 | C7H4Cl2N2S | 219.083 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo A14309.03
también denominado A14309-03
Precio (USD)
-
Cantidad:
1 g
Pedido a granel o personalizado
Identificadores químicos
CAS19462-98-7
IUPAC Name5,6-dichloro-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Molecular FormulaC7H4Cl2N2S
InChI KeyAFDOMGKBKBKUHB-UHFFFAOYSA-N
SMILESClC1=C(Cl)C=C2NC(=S)NC2=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Pale yellow to pale brown
Assay (HPLC)≥97.5%
FormCrystals or powder or crystalline powder
It is employed in the synthesis of 2?,3?-dideoxy and 2?,3?-unsaturated ribofuranonucleosides as potential antiviral agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
Se emplea en la síntesis de 2′,3′-dideoxi y ribofuranonucleósidos 2′,3′-insaturados como agentes antivirales potenciales.

Solubilidad
Insoluble en agua.

Notas
Almacenar lejos de agentes oxidantes. Mantenga el recipiente bien cerrado en un lugar fresco, seco y bien ventilado.
RUO – Research Use Only

General References:

  1. Edward S. Lazer, et.al. Benzimidazole derivatives with atypical antiinflammatory activity.J. Med. Chem.,1987,30(4), 726-729.
  2. Christophe Mathé, et al. Nucleoside analogues. Synthesis of 2',3'-dideoxy and 2',3'-unsaturated ribofuranonucleosides of 5,6-dichloro-2-mercaptobenzimidazole as potential antiviral agents.J. Chem. Soc., Perkin Trans.,1994,1(8), 1019-1024.