3-(2-Methoxyphenyl)propionic acid, 98+%
3-(2-Methoxyphenyl)propionic acid, 98+%
3-(2-Methoxyphenyl)propionic acid, 98+%
Thermo Scientific Chemicals

3-(2-Methoxyphenyl)propionic acid, 98+%

CAS: 6342-77-4 | C10H12O3 | 180.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14314.06
also known as A14314-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6342-77-4
IUPAC Name3-(2-methoxyphenyl)propanoic acid
Molecular FormulaC10H12O3
InChI KeyXSZSNLOPIWWFHS-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)83.5-89.5?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only