N-(Cyanomethyl)pyridinium chloride, 97%
N-(Cyanomethyl)pyridinium chloride, 97%
N-(Cyanomethyl)pyridinium chloride, 97%
Thermo Scientific Chemicals

N-(Cyanomethyl)pyridinium chloride, 97%

CAS: 17281-59-3 | C7H7ClN2 | 154.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A14327.18
also known as A14327-18
Price (EUR)
131,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
131,00
Each
Chemical Identifiers
CAS17281-59-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Melting Point (clear melt)163.5-169.5?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only