2',4',6'-Trimethoxyacetophenone, 98%
2',4',6'-Trimethoxyacetophenone, 98%
2',4',6'-Trimethoxyacetophenone, 98%
Thermo Scientific Chemicals

2',4',6'-Trimethoxyacetophenone, 98%

CAS: 832-58-6 | C11H14O4 | 210.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14328.14
also known as A14328-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS832-58-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)100.0-106.0?C
2',4',6'-Trimethoxyacetophenone is used in the production of 4,2',4',6'-tetramethoxy-chalcone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,4′,6′-Trimethoxyacetophenone is used in the production of 4,2′,4′,6′-tetramethoxy-chalcone.

Solubility
Soluble in chloroform.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhang, J.; Liu, M.; Cui, W.; Yang, J.; Yang, B. Total synthesis of luteolin. J. Chem. Res. 2014, 38 (1), 60-61.
  2. Nasir Abbas Bukhari, S.; Jantan, I.; Jasamai, M. Anti-Inflammatory Trends of 1, 3-Diphenyl-2-propen-1-one Derivatives. Mini Rev Med Chem 2013, 13 (1), 87-94.