Adamantane-1-carbonitrile, 97%
Adamantane-1-carbonitrile, 97%
Adamantane-1-carbonitrile, 97%
Adamantane-1-carbonitrile, 97%
Thermo Scientific Chemicals

Adamantane-1-carbonitrile, 97%

CAS: 23074-42-2 | C11H15N | 161.248 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A14352.14
also known as A14352-14
Price (EUR)
347,65
Online Exclusive
409,00
Save 61,35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
347,65
Online Exclusive
409,00
Save 61,35 (15%)
Each
Add to cart
Chemical Identifiers
CAS23074-42-2
IUPAC Nameadamantane-1-carbonitrile
Molecular FormulaC11H15N
InChI KeyFQFZASRJFRAEIH-UHFFFAOYSA-N
SMILESN#CC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)183-198?C
Appearance (Color)White to pale cream or pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only