2-Mercaptoethylamine hydrochloride, 97+%
2-Mercaptoethylamine hydrochloride, 97+%
2-Mercaptoethylamine hydrochloride, 97+%
2-Mercaptoethylamine hydrochloride, 97+%
Thermo Scientific Chemicals

2-Mercaptoethylamine hydrochloride, 97+%

CAS: 156-57-0 | C2H8ClNS | 113.60 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
產品號碼 A14377.22
亦稱為 A14377-22
價格 (TWD)
-
申請報價
Quantity:
100 g
Request bulk or custom format
化學識別
CAS156-57-0
IUPAC Namehydrogen 2-aminoethane-1-thiol chloride
Molecular FormulaC2H8ClNS
InChI KeyOGMADIBCHLQMIP-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCS
檢視更多
規格Specification Sheet規格表
FormCrystals or powder or crystalline powder or chunks or lumps or fused solid
Identification (FTIR)Conforms
Appearance (Color)White
Assay (Redox Titration)≥97.0 to ≤103.0%
Melting Point (clear melt)63.0-70.0?C
An inhibitor of DMBA-induced mammary tumors2-Mercaptoethylamine hydrochloride acts as a precursor in taurine biosynthesis and component of coenzyme A. It is involved in the preparation of active pharmaceutical ingredients like ranitidine and nizatidine. It acts as an antidote for acetaminophen poisoning. Further, it is used in the oral treatment of nephropathic cystinosis, radiation sickness and disorders of cysteine excretion. In addition to this, it serves as an antioxidant and an inhibitor of 7,12-Dimethylbenz[a]anthracene (DMBA)-induced tumors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An inhibitor of DMBA-induced mammary tumors2-Mercaptoethylamine hydrochloride acts as a precursor in taurine biosynthesis and component of coenzyme A. It is involved in the preparation of active pharmaceutical ingredients like ranitidine and nizatidine. It acts as an antidote for acetaminophen poisoning. Further, it is used in the oral treatment of nephropathic cystinosis, radiation sickness and disorders of cysteine excretion. In addition to this, it serves as an antioxidant and an inhibitor of 7,12-Dimethylbenz[a]anthracene (DMBA)-induced tumors.

Solubility
Soluble in water, ethanol, methanol and dimethyl sulfoxide.

Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. For selective protection of nitrogen by silylation, see: Synthesis, 924 (1980).
  2. Bhattacharjee, S.; Lee, Y. R.; Ahn, W. S. Post-synthesis functionalization of a zeolitic imidazolate structure ZIF-90: a study on removal of Hg(II) from water and epoxidation of alkenes. CrystEngComm 2015, 17 (12), 2575-2582.
  3. Yuan, Y. Y.; Du, J. Z.; Wang, J. Two consecutive click reactions as a general route to functional cyclic polyesters. Chem. Commun. 2012, 48 (4), 570-572.