2-Bromoterephthalic acid, 97%
2-Bromoterephthalic acid, 97%
2-Bromoterephthalic acid, 97%
Thermo Scientific Chemicals

2-Bromoterephthalic acid, 97%

CAS: 586-35-6 | C8H5BrO4 | 245.028 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14393.06
also known as A14393-06
Price (EUR)
59,80
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
59,80
Each
Chemical Identifiers
CAS586-35-6
IUPAC Name2-bromobenzene-1,4-dicarboxylic acid
Molecular FormulaC8H5BrO4
InChI KeyQPBGNSFASPVGTP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C(O)=O)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)295.0-304.0?C
Assay (HPLC)≥96.0%
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormPowder or granules
2-Bromoterephthalic acid is used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoterephthalic acid is used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Pradip K. Bhowmik, Haesook Han. Fully aromatic liquid-crystalline polyesters of phenyl-substituted 4,4'-biphenols and 1,1'-binaphthyl-4,4'-diol with either 2-bromoterephthalic acid or 2-phenylterephthalic acid. Macromolecules. 1993, 26 (20), 5287-5294.
  2. Jung Il Jin.; E Joon Choi.; Byung Wook Jo. Properties of poly(p-phenylene terephthalates) prepared from 2-nitro- and 2-bromoterephthalic acids and substituted hydroquinones. Macromolecules. 1987, 20 (5), 934-939.