Phenyltrimethylammonium iodide, 99%
Phenyltrimethylammonium iodide, 99%
Phenyltrimethylammonium iodide, 99%
Phenyltrimethylammonium iodide, 99%
Thermo Scientific Chemicals

Phenyltrimethylammonium iodide, 99%

CAS: 98-04-4 | C9H14IN | 263.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14402.22
also known as A14402-22
Price (EUR)
90,30
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
90,30
Each
Add to cart
Chemical Identifiers
CAS98-04-4
IUPAC NameN,N,N-trimethylanilinium iodide
Molecular FormulaC9H14IN
InChI KeyKKLAORVGAKUOPZ-UHFFFAOYSA-M
SMILES[I-].C[N+](C)(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Iodide)≥98.5 to ≤101.5%
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water and methanol.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Robert O. Hutchins.; Duraisamy Kandasamy.; Frank DuxIII.; Cynthia A. Maryanoff.; David Rotstein.; Barry Goldsmith.; William Burgoyne.; Frank Cistone.; Joseph Dalessandro.; Joseph Puglis. Nucleophilic borohydride: selective reductive displacement of halides, sulfonate esters, tertiary amines, and N,N-disulfonimides with borohydride reagents in polar aprotic solvents. J. Org. Chem. 1978, 43 (11), 2259-2267.
  2. A. Pass, A.I.C.; A. M. Ward, D.Sc., A.I.C. The determination of cadmium in the presence of zinc, in spelter and in zinc ores. Analyst. 1933, 58 667-672.
  3. Reagent for Cd.