Thermo Scientific Chemicals

Methyl 3-hydroxybenzoate, 99%, Thermo Scientific Chemicals

Catalog number: A14425.22
100 g, Each
Thermo Scientific Chemicals

Methyl 3-hydroxybenzoate, 99%, Thermo Scientific Chemicals

Catalog number: A14425.22
100 g, Each
Quantity
Catalog number: A14425.22
also known as A14425-22
Price (USD)
Price: 161.00
Online price: 138.65
Your price:
Quantity
-

Chemical Identifiers

CAS
19438-10-9
IUPAC Name
methyl 3-hydroxybenzoate
Molecular Formula
C8H8O3
InChI Key
YKUCHDXIBAQWSF-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC(O)=C1
Appearance (Color)
White to cream to pale gray
Assay (GC)
≥98.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
67.0-75.0°C

Description

Methyl 3-hydroxybenzoate is used in the preparation of 3-hydroxybenzene-1,2-dicarbaldehyde and O-methyl O-[3-methyl-4-(methylthio)phenyl]O-(3-methylcarboxyphenyl) phosphorothioate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-hydroxybenzoate is used in the preparation of 3-hydroxybenzene-1,2-dicarbaldehyde and O-methyl O-[3-methyl-4-(methylthio)phenyl]O-(3-methylcarboxyphenyl) phosphorothioate.

Solubility
Soluble in chloroform, dimetyl Sulfoxide,ethanol and methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text