Indol, 99 %, Thermo Scientific Chemicals
Indol, 99 %, Thermo Scientific Chemicals
Indol, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indol, 99 %, Thermo Scientific Chemicals

CAS: 120-72-9 | C8H7N | 117.151 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
50 g
250 g
1000 g
Número de catálogo A14427.18
también denominado A14427-18
Precio (EUR)
50,65
Precio exclusive compra web
56,40
Ahorro 5,75 (10%)
Each
Cantidad:
50 g
Pedido a granel o personalizado
Precio (EUR)
50,65
Precio exclusive compra web
56,40
Ahorro 5,75 (10%)
Each
Identificadores químicos
CAS120-72-9
IUPAC Name1H-indole
Molecular FormulaC8H7N
InChI KeySIKJAQJRHWYJAI-UHFFFAOYSA-N
SMILESN1C=CC2=CC=CC=C12
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
FormCrystals or powder or crystalline powder or flakes
Appearance (Color)White to pale cream
Assay (GC)≥98.5% (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Water Content (Karl Fischer Titration)≤1% (non-U.S. specification)
Ver más
Indole is used in the preparation of gramine, which is obtained by Mannich reaction dimethylamine and formaldehyde. It is used as a starting material for the preparation of oxindole and indole-3-carboxaldehyde. In Diels-Alder reaction, it acts as dienophile and reacts with 2-aminofuran to get strychnine. It is the active component of jasmine oil, neroli oil and orange-blossom oil used in the perfume industry. Further, it is involved in the preparation of synthetic jasmine oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El indol se utiliza en la preparación de la gramina, que se obtiene por dimetilamina y formaldehído en reacción de Mannich. Se utiliza como material de partida para la preparación de oxindol e indol-3-carboxaldehído. En la reacción de Diels-Alder, actúa como dienófilo y reacciona con 2-aminofurano para obtener estricnina. Es el componente activo del aceite de jazmín, aceite de nerolí y aceite de flor de naranjo, utilizados en la industria del perfume. Además, participa en la preparación de aceite de jazmín sintético.

Solubilidad
Soluble en metanol y agua.

Notas
Sensible a la luz y al aire. Almacenar en lugar fresco. Incompatible con agentes oxidantes fuertes, hierro y sales de hierro. Mantenga el recipiente bien cerrado en un lugar seco y bien ventilado.
RUO – Research Use Only

General References:

  1. Lithiation of N-protected indoles usually occurs at the 2-position, e.g. benzenesulfonation of the 1-lithio derivative, 2-lithiation with LDA and treatment with pyridine-3,4-dicarboxylic anhydride. This sequence has been reported in a synthetic route to ellipticene and olivacine: J. Org. Chem., 57, 5891 (1992).
  2. Regioselective synthesis of 3-substituted indoles has been described using a sequence of N-silylation, 3-bromination of the 1-TBDMS detivative with NBS, 3-lithiation, treatment with an electrophile to introduce the 3-substituent, and desilylation with TBAF. Good overall yields are obtained for a range of electrophiles: J. Org. Chem., 59, 10 (1994); Org. Synth. Coll., 9, 417 (1998).
  3. Treatment with EtMgI followed by ZnCl2 leads to an N-zinc derivative. This undergoes acylation at the 3-position: Tetrahedron, 46, 6061 (1990).
  4. Halogenation in the 2-position has been effected via the Li 1-carbamate and subsequent 2-lithiation: J. Org. Chem., 57, 2495 (1992):
  5. Treatment of the carbamate with Tri-n-butyl tin chloride, A10746, followed by Stille coupling enables synthesis of 2-aryl- and 2-vinylindoles: J. Org. Chem., 60, 6218 (1995).
  6. Johnston, A. J.; Zhang, Y. R.; Busch, S.; Pardo, L. C.; Imberti, S.; McLain, S. E. Amphipathic Solvation of Indole: Implications for the Role of Tryptophan in Membrane Proteins. J. Phys. Chem. B 2015, 119 (19), 5979-5987.
  7. Yang, P.; Pang, M.; Shen, W.; Li, M.; He, R. Vibronic analysis of the 1Lb ↔ S0 transitions of indole and 3-methylindole: The influence of anharmonic, Duschinsky, and Herzberg-Teller contributions. J. Mol. Spectrosc. 2015, 313, 40-48.