2,6-Dihydroxybenzoic acid, 98%
2,6-Dihydroxybenzoic acid, 98%
2,6-Dihydroxybenzoic acid, 98%
Thermo Scientific Chemicals

2,6-Dihydroxybenzoic acid, 98%

CAS: 303-07-1 | C7H6O4 | 154.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14485.14
also known as A14485-14
Price (EUR)
45,40
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
45,40
Each
Chemical Identifiers
CAS303-07-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only