2-Furoylacetonitrile, 97%
2-Furoylacetonitrile, 97%
2-Furoylacetonitrile, 97%
Thermo Scientific Chemicals

2-Furoylacetonitrile, 97%

CAS: 31909-58-7 | C7H5NO2 | 135.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A14497.14
also known as A14497-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS31909-58-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)73.0-83.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only