Thermo Scientific Chemicals

2-Methylanthraquinone, 97%, Thermo Scientific Chemicals

Catalog number: A14502.30
250 g, Each
Thermo Scientific Chemicals

2-Methylanthraquinone, 97%, Thermo Scientific Chemicals

Catalog number: A14502.30
250 g, Each
Quantity
Catalog number: A14502.30
also known as A14502-30
Price (USD)
Quantity
-

Chemical Identifiers

CAS
84-54-8
IUPAC Name
2-methyl-9,10-dihydroanthracene-9,10-dione
Molecular Formula
C15H10O2
InChI Key
NJWGQARXZDRHCD-UHFFFAOYSA-N
SMILES
CC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
Appearance (Color)
Yellow
Melting Point (clear melt)
171.0-180.0?C
Form
Powder
Assay (GC)
≥96.0%

Description

2-Methylanthraquinone is used as a pharmaceutical intermediate. It is used in smog dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylanthraquinone is used as a pharmaceutical intermediate. It is used in smog dyes.

Solubility
Soluble in water 1.234 mg/L @ 25°C.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text