N-(4-Bromobutyl)phthalimide, 96%
N-(4-Bromobutyl)phthalimide, 96%
N-(4-Bromobutyl)phthalimide, 96%
N-(4-Bromobutyl)phthalimide, 96%
Thermo Scientific Chemicals

N-(4-Bromobutyl)phthalimide, 96%

CAS: 5394-18-3 | C12H12BrNO2 | 282.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14517.06
also known as A14517-06
Price (EUR)
56,30
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
56,30
Each
Add to cart
Chemical Identifiers
CAS5394-18-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (GC)≥95.0%
FormPowder
N-(4-Bromobutyl)phthalimide is used in organic synthesis and the production of pharmaceutical. It can react with 1-phenyl-piperazine to get N-[4-(4-phenyl-piperazin-1-yl)-butyl]-phthalimide. It is a useful synthesis reagent used to synthesize B-cyclodextrin derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(4-Bromobutyl)phthalimide is used in organic synthesis and the production of pharmaceutical. It can react with 1-phenyl-piperazine to get N-[4-(4-phenyl-piperazin-1-yl)-butyl]-phthalimide. It is a useful synthesis reagent used to synthesize B-cyclodextrin derivatives.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store at room temperature. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. S. Tomasi; M. Le Roch; J. Renault; J. C. Corbel and P. Uriac. N-alkylation of N1-mesitylenesulphonylputrescine with N-(4-bromobutyl)phthalimide: A Parallel Approach Using Multipin Solid-phase Synthesis. Pharmacy and Pharmacology Communications. 2000, 6 (4), 155-159.
  2. Richard A. Glennon; Noreen A. Naiman; M. Edward Pierson; J. Doyle Smith; Abd M. Ismaiel; Milt Titeler; Robert A. Lyon. N-(phthalimidoalkyl) derivatives of serotonergic agents: a common interaction at 5-HT1A serotonin binding sites?. J. Med. Chem. 1989, 32 (8), 1921-1926.