Thermo Scientific Chemicals

Potassium L-tartrate hemihydrate, 99%, Thermo Scientific Chemicals

Catalog number: A14556.0I
5000 g, Each
Thermo Scientific Chemicals

Potassium L-tartrate hemihydrate, 99%, Thermo Scientific Chemicals

Catalog number: A14556.0I
5000 g, Each
Quantity
Catalog number: A14556.0I
also known as A14556-0I
Price (USD)
Price: 533.00
Online price: 460.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6100-19-2
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White

Description

Potassium L-tartrate hemihydrate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium L-tartrate hemihydrate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text