Pyridine-3,4-dicarboxylic acid, 98%
Pyridine-3,4-dicarboxylic acid, 98%
Pyridine-3,4-dicarboxylic acid, 98%
Pyridine-3,4-dicarboxylic acid, 98%
Thermo Scientific Chemicals

Pyridine-3,4-dicarboxylic acid, 98%

CAS: 490-11-9 | C7H5NO4 | 167.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A14580.18
also known as A14580-18
Price (EUR)
166,00
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
166,00
Each
Add to cart
Chemical Identifiers
CAS490-11-9
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Aqueous acid-base Titration)>97.5 to <102.5%
Water Content (Karl Fischer Titration)<1.0%
Appearance (Color)White to pale brown
Assay (Silylated GC)>97.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only