6-Chloro-1-hexanol, 97%
6-Chloro-1-hexanol, 97%
6-Chloro-1-hexanol, 97%
Thermo Scientific Chemicals

6-Chloro-1-hexanol, 97%

CAS: 2009-83-8 | C6H13ClO | 136.619 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14582.14
also known as A14582-14
Price (EUR)
59,50
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
59,50
Each
Chemical Identifiers
CAS2009-83-8
IUPAC Name6-chlorohexan-1-ol
Molecular FormulaC6H13ClO
InChI KeyJNTPTNNCGDAGEJ-UHFFFAOYSA-N
SMILESOCCCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.4535-1.4585 @ 20?C
6-Chloro-1-hexanol is used in the preparation of its corresponding bromide by reaction with sodium bromide using N,N-dimethylformamide and dibromomethane (2:1) as a solvent. It is utilized in the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloro-1-hexanol is used in the preparation of its corresponding bromide by reaction with sodium bromide using N,N-dimethylformamide and dibromomethane (2:1) as a solvent. It is utilized in the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.

Solubility
Soluble in dimethylformamide.

Notes
Incompatible with strong oxidizing agents, strong bases, powdered metal salts, strong reducing agents, acid anhydrides and acid chlorides.
RUO – Research Use Only

General References:

  1. Cimen, D.; Yildirim, E.; Caykara, T. Synthesis of dual-functional poly(6-azidohexylmethacrylate) brushes by a RAFT agent carrying carboxylic acid end groups. J. Polym. Sci., Part A: Polym. Chem. 2015, 53 (14), 1696-1706.
  2. Xu, L.; Liu, L.; Liu, F.; Cai, H.; Zhang, W. Porphyrin-containing amphiphilic block copolymers for photodynamic therapy. Polym. Chem. 2015, 6 (15), 2945-2954.