Thermo Scientific Chemicals

6-Chloro-1-hexanol, 97%, Thermo Scientific Chemicals

Catalog number: A14582.22
100 g, Each
Thermo Scientific Chemicals

6-Chloro-1-hexanol, 97%, Thermo Scientific Chemicals

Catalog number: A14582.22
100 g, Each
Quantity
Catalog number: A14582.22
also known as A14582-22
Price (USD)
Price: 183.00
Online price: 174.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2009-83-8
IUPAC Name
6-chlorohexan-1-ol
Molecular Formula
C6H13ClO
InChI Key
JNTPTNNCGDAGEJ-UHFFFAOYSA-N
SMILES
OCCCCCCCl
Identification (FTIR)
Conforms
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Refractive Index
1.4535-1.4585 @ 20?C

Description

6-Chloro-1-hexanol is used in the preparation of its corresponding bromide by reaction with sodium bromide using N,N-dimethylformamide and dibromomethane (2:1) as a solvent. It is utilized in the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloro-1-hexanol is used in the preparation of its corresponding bromide by reaction with sodium bromide using N,N-dimethylformamide and dibromomethane (2:1) as a solvent. It is utilized in the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.

Solubility
Soluble in dimethylformamide.

Notes
Incompatible with strong oxidizing agents, strong bases, powdered metal salts, strong reducing agents, acid anhydrides and acid chlorides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text