Thermo Scientific Chemicals

Glutaric acid, 99%, Thermo Scientific Chemicals

Catalog number: A14595.36
500 g, Each
Thermo Scientific Chemicals

Glutaric acid, 99%, Thermo Scientific Chemicals

Catalog number: A14595.36
500 g, Each
Quantity
Catalog number: A14595.36
also known as A14595-36
Price (USD)
Price: 219.00
Online price: 186.65
Your price:
Quantity
-

Chemical Identifiers

CAS
110-94-1
IUPAC Name
pentanedioic acid
Molecular Formula
C5H8O4
InChI Key
JFCQEDHGNNZCLN-UHFFFAOYSA-N
SMILES
OC(=O)CCCC(O)=O
Assay (Aqueous acid-base Titration)
≥98.5%
Appearance (Color)
White to pale cream
Assay (Silylated GC)
≥98.5%
Melting Point
95.0-101.0?C
Form
Crystals or powder or crystalline powder

Description

Glutaric acid is used as the raw material for organic synthesis, pharmaceutical intermediate and synthetic resin. It serves as a precursor in the production of polyester polyols, polyamides, ester plasticizers and corrosion inhibitors. It is useful to decrease polymer elasticity and in the synthesis surfactants and metal finishing compounds. It acts as an intermediate during the catabolism of lysine in mammals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaric acid is used as the raw material for organic synthesis, pharmaceutical intermediate and synthetic resin. It serves as a precursor in the production of polyester polyols, polyamides, ester plasticizers and corrosion inhibitors. It is useful to decrease polymer elasticity and in the synthesis surfactants and metal finishing compounds. It acts as an intermediate during the catabolism of lysine in mammals.

Solubility
Soluble in water, alcohol, benzene and chloroform. Slightly soluble in petroleum ether.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text