How many times can I passage the Human Umbilical Vein Endothelial Cells (HUVECs) before performing the tube formation assay using the Angiogenesis Starter Kit?
For optimal performance, we recommend using HUVECs passaged less than 4 times.