2,4-Difluorobenzenesulfonyl chloride, 98%
2,4-Difluorobenzenesulfonyl chloride, 98%
2,4-Difluorobenzenesulfonyl chloride, 98%
2,4-Difluorobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

2,4-Difluorobenzenesulfonyl chloride, 98%

CAS: 13918-92-8 | C6H3ClF2O2S | 212.595 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14639.22
also known as A14639-22
Price (TWD)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS13918-92-8
IUPAC Name2,4-difluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H3ClF2O2S
InChI KeyFJSAJUXIHJIAMD-UHFFFAOYSA-N
SMILESFC1=CC(F)=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5155-1.5195 @ 20?C
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only