Thermo Scientific Chemicals

6-Acetyl-1,2,3,4-tetrahydronaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: A14666.22
100 g, Each
Thermo Scientific Chemicals

6-Acetyl-1,2,3,4-tetrahydronaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: A14666.22
100 g, Each
Quantity
Catalog number: A14666.22
also known as A14666-22
Price (USD)
Price: 433.00
Online price: 374.65
Your price:
Quantity
-

Chemical Identifiers

CAS
774-55-0
IUPAC Name
1-(5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Molecular Formula
C12H14O
InChI Key
VEPUKHYQNXSSKV-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C2CCCCC2=C1
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.5590-1.5640 @ 20?C
Form
Liquid
Assay (GC)
≥96.0%

Description

6-Acetyl-1,2,3,4-tetrahydronaphthalene, is used asa novel methyl anthranilate yet extremely stable and resistant to discoloration. It is used in liquid detergent, concentrated fabric softner, alcoholic fine fragrances, antiperspirants. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Acetyl-1,2,3,4-tetrahydronaphthalene, is used asa novel methyl anthranilate yet extremely stable and resistant to discoloration. It is used in liquid detergent, concentrated fabric softner, alcoholic fine fragrances, antiperspirants. It is also used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Product is sensitive to light and moisture. Handle and store under inert gas. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text