3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 97%
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 97%
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 97%
Thermo Scientific Chemicals

3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 97%

CAS: 88-26-6 | C15H24O2 | 236.355 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14709.22
also known as A14709-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS88-26-6
IUPAC Name2,6-di-tert-butyl-4-(hydroxymethyl)phenol
Molecular FormulaC15H24O2
InChI KeyHNURKXXMYARGAY-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(CO)=CC(=C1O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)135.0-144.0?C
Appearance (Color)Pale yellow to yellow
FormPowder
Assay (Silylated GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only