Thermo Scientific Chemicals

Hexanophenone, 98%, Thermo Scientific Chemicals

Catalog number: A14712.22
100 g, Each
Thermo Scientific Chemicals

Hexanophenone, 98%, Thermo Scientific Chemicals

Catalog number: A14712.22
100 g, Each
Quantity
Catalog number: A14712.22
also known as A14712-22
Price (USD)
Price: 283.00
Online price: 244.65
Your price:
Quantity
-

Chemical Identifiers

CAS
942-92-7
IUPAC Name
1-phenylhexan-1-one
Molecular Formula
C12H16O
InChI Key
MAHPVQDVMLWUAG-UHFFFAOYSA-N
SMILES
CCCCCC(=O)C1=CC=CC=C1
Appearance (Color)
Colorless to white to pale yellow
Form
Fused solid or clear liquid as melt
Assay (GC)
≥97.5%
Refractive Index
1.5090-1.5130 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text