1-Bromooctane, 98+%
1-Bromooctane, 98+%
1-Bromooctane, 98+%
Thermo Scientific Chemicals

1-Bromooctane, 98+%

CAS: 111-83-1 | C8H17Br | 193.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A14722.0I
also known as A14722-0I
Price (EUR)
485,00
Each
Quantity:
5000 g
Request bulk or custom format
Price (EUR)
485,00
Each
Chemical Identifiers
CAS111-83-1
IUPAC Name1-bromooctane
Molecular FormulaC8H17Br
InChI KeyVMKOFRJSULQZRM-UHFFFAOYSA-N
SMILESCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index1.4505-1.4545 @ 20?C
FormLiquid
1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Iqbal, Z.; Wu, W. Q.; Huang, Z. S.; Wang, L.; Kuang, D. B.; Meier, H.; Cao, D. Trilateral pi-conjugation extensions of phenothiazine-based dyes enhance the photovoltaic performance of the dye-sensitized solar cells. Dyes Pigm. 2016, 124, 63-71.
  2. Sun, T.; Qi, M.; Fu, R. Perhydroxylcucurbit[6]uril as a highly selective gas chromatographic stationary phase for analytes of wide-ranging polarity. J. Sep. Sci. 2015, 38 (5), 821-824.