Thermo Scientific Chemicals

Diethylene glycol, 99%, Thermo Scientific Chemicals

Catalog number: A14728.30
250 g, Each
Thermo Scientific Chemicals

Diethylene glycol, 99%, Thermo Scientific Chemicals

Catalog number: A14728.30
250 g, Each
Quantity
Catalog number: A14728.30
also known as A14728-30
Price (USD)
Price: 22.50
Online price: 19.65
Your price:
Quantity
-

Chemical Identifiers

CAS
111-46-6
IUPAC Name
2-(2-hydroxyethoxy)ethan-1-ol
Molecular Formula
C4H10O3
InChI Key
MTHSVFCYNBDYFN-UHFFFAOYSA-N
SMILES
OCCOCCO
Assay (GC)
≥98.5% (UK Source Material)
Assay from Supplier's CofA
≥98.5% (US Sourced Material)
Comment
Material Sourced in UK and US
Identification (FTIR)
Conforms (UK Sourced Material)
Water Content (Karl Fischer Titration)
≤0.5%

Description

Diethylene glycol is used as an important industrial solvent due to its characteristic properties like high boiling point, high viscosity and low vapor loss. It is widely used in the manufacture of unsaturated polyester resins, polyurethanes, nitrocellulose, resins and plasticizers. It plays an important role as a building block in the preparation of morpholine and 1,4-dioxane. It finds application as a heat transfer fluid in the chemical industry due to its low freezing point. As a lubricant, it is used in glass-grinding aids, fiber-finish component and cement-grinding aids. It is also used in natural gas to remove water and other impurities. It is an active component of brake fluid, artificial fog solutions and wallpaper strippers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol is used as an important industrial solvent due to its characteristic properties like high boiling point, high viscosity and low vapor loss. It is widely used in the manufacture of unsaturated polyester resins, polyurethanes, nitrocellulose, resins and plasticizers. It plays an important role as a building block in the preparation of morpholine and 1,4-dioxane. It finds application as a heat transfer fluid in the chemical industry due to its low freezing point. As a lubricant, it is used in glass-grinding aids, fiber-finish component and cement-grinding aids. It is also used in natural gas to remove water and other impurities. It is an active component of brake fluid, artificial fog solutions and wallpaper strippers.

Solubility
Miscible with water, alcohol, ether, acetone, and ethylene glycol.

Notes
Hygroscopic. Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and zinc.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text