2-Metilbenzofenona, + 98 %, Thermo Scientific Chemicals
2-Metilbenzofenona, + 98 %, Thermo Scientific Chemicals
2-Metilbenzofenona, + 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Metilbenzofenona, + 98 %, Thermo Scientific Chemicals

CAS: 131-58-8 | C14H12O | 196.249 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
10 g
50 g
250 g
Número de catálogo A14779.30
también denominado A14779-30
Precio (USD)
-
Cantidad:
250 g
Pedido a granel o personalizado
Identificadores químicos
CAS131-58-8
IUPAC Name(2-methylphenyl)(phenyl)methanone
Molecular FormulaC14H12O
InChI KeyCKGKXGQVRVAKEA-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C(=O)C1=CC=CC=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
FormLiquid
Refractive Index1.5930-1.5970 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.0%
2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
La 2-metilbenzofenona se utiliza como una importante materia prima e intermediaria en síntesis orgánica, productos farmacéuticos, productos agroquímicos y colorantes.

Solubilidad
Insoluble en agua.

Notas
Almacenar en un lugar fresco. Mantenga el recipiente bien cerrado en un lugar seco y bien ventilado. Mantener alejado de agentes oxidantes fuertes. Estable bajo temperatura y presión normales.
RUO – Research Use Only

General References:

  1. PK Das.; JC Scaiano. The formation of two photoenols via a common pathway in the photochemistry of 2-methylbenzophenone. Journal of Photochemistry.1980, 12 (1), 85-90.
  2. T Nakayama.; K Hamanoue.; T Hidaka.; M Okamoto. Photoenolization of 2-methylbenzophenone studied by picosecond and nanosecond laser spectroscopy. Journal of Photochemistry.1980, 24 (1), 71-78.
  3. For a review of the Elbs reaction, in which 2-alkylbenzophenones are cyclized at high temperatures to anthracenes, see: Org. React., 1, 129 (1942).