2-(Trifluoromethyl)phenylhydrazine is used as an intermediate in organic syntheses. It is also ued as a fine chemical intermediate, GC derivatizing agent, coupling reagent and N- protecting agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
La 2-(trifluorometil)fenilhidracina se utiliza como intermediario en la síntesis orgánica. También se usa como un químico intermedio fino, agente de derivatización de GC, reactivo de acoplamiento y agente N-protector.
Solubilidad
Insoluble en agua.
Notas
Almacenar en lugar fresco. Mantenga el recipiente bien cerrado en un lugar seco y bien ventilado. Mantener alejado de agentes oxidantes fuertes.
RUO – Research Use Only
General References:
- C Kunick.; C Schultz.; T Lemcke.; DW Zaharevitz. 2-Substituted paullones: CDK1/cyclin B-inhibiting property and in vitro antiproliferative activity. Bioorganic & Medicinal Chemistry Letters. 1999, 10 (6), 567-569.
- H Jiang.; Y Wang.; W Wan.; J Hao. p-TsOH promoted Fischer indole synthesis of multi-substituted 2-trifluoromethyl indole derivatives. Tetrahedron. 2010, 10 (6), 567-569.