Decanoic acid, 99%
Decanoic acid, 99%
Decanoic acid, 99%
Thermo Scientific Chemicals

Decanoic acid, 99%

CAS: 334-48-5 | C10H20O2 | 172.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A14788.0B
also known as A14788-0B
Price (TWD)
-
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS334-48-5
IUPAC Namedecanoic acid
Molecular FormulaC10H20O2
InChI KeyGHVNFZFCNZKVNT-UHFFFAOYSA-N
SMILESCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or chunks or fused solid
Identification (FTIR)Conforms
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (Silylated GC)≥98.5%
View more
Decanoic acid is used in perfumes, resins, lubricants, greases, dyes, plastics, pharmaceuticals and food additives. It is involved in the preparation of wetting agents and sodium deconate, which is an active component in soap. It is also used as a plasticizer and as an absorption enhancer in guinea pig skin experiments. Further, it is employed in the generation of esters for artificial fruit flavors. In addition to this, it is used in agricultural chemicals, lubricant additives and surface active agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decanoic acid is used in perfumes, resins, lubricants, greases, dyes, plastics, pharmaceuticals and food additives. It is involved in the preparation of wetting agents and sodium deconate, which is an active component in soap. It is also used as a plasticizer and as an absorption enhancer in guinea pig skin experiments. Further, it is employed in the generation of esters for artificial fruit flavors. In addition to this, it is used in agricultural chemicals, lubricant additives and surface active agents.

Solubility
Soluble in acetone, benzene, chloroform, carbon disulfide, ethyl ether, ethanol, dilute nitric acid and most organic solvents. Insoluble in water.

Notes
Incompatible with bases, reducing agents and oxidizing agents.
RUO – Research Use Only

General References:

  1. Wang, J.; Zhang, L.; Xin, D.; Yang, Y. Dispersive Micro-Solid-Phase Extraction Based on Decanoic Acid Coated-Fe3O4 Nanoparticles for HPLC Analysis of Phthalate Esters in Liquor Samples. J. Food Sci. 2015, 80 (11), C2452-C2458.
  2. Ikari, K.; Sakuma, Y.; Jimbo, T.; Kodama, A.; Imai, M.; Monnard, P. A.; Rasmussen, S. Dynamics of fatty acid vesicles in response to pH stimuli. Soft Matter 2015, 11 (31), 6327-6334.